missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PCR Plate, 384-well, raised chimney

Product Code. 10374273
Change view
Click to view available options
Quantity:
100 plates
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10374273 100 plates Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10374273 Supplier Thermo Scientific™ Supplier No. AB0937

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform post-PCR sequencing and wash steps in the larger wells of this 384-well PCR plate with a 40 μL well volume.

Extra Volume 384-well full-skirted plate permits enables post-PCR applications such as enzymatic digestions and ethanol precipitation to be carried out in the wells due to the increased well volume.

Highlights

  • Tall Chimney well design
  • Increased well volume accommodates sequencing and wash steps
  • Working well volume: 40 µL
  • Cut corner: A24
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 100 Plate
Color Clear
Product Type PCR Microplate
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Skirt Style Skirted
Material Polypropylene
Volume (Metric) Well 40 μL
No. of Wells 384
Volume (Metric) Working 25 μL
Quantity 100 plates
Reaction Speed Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.