missing translation for 'onlineSavingsMsg'
Learn More

Motic™ 360° Rotatable Analyzer

Product Code. 17109378
Click to view available options
For Use With (Equipment):
Motic TEC POL Microscopes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17109378

Brand: Motic™ 1101001100643

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Completely rotatable analyzer for use with TEC POL Microscopes.

  • For use with Motic™ Panthera TEC POL Microscopes
  • Rotates 360°

Specifications

Type Microscope Analyzer
Color Black
For Use With (Application) Controls Strength of Light
For Use With (Equipment) Motic TEC POL Microscopes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.