missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Sequencing Standard, BigDye™ Terminator v1.1 Dx

Product Code. 15819866 Shop All Applied Biosystems Products
Click to view available options
Quantity:
4 Tubes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15819866

Brand: Applied Biosystems™ 4462113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

This kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye® Terminator v1.1. Each tube can perform one spectral calibration/performance check and/or one control sequencing run on the Applied Biosystems™ Dx Series Genetic Analyzers

Sufficient DNA is included to perform one spectral calibration/performance check or one control sequencing run on Applied Biosystems Dx genetic analyzers.

Sequencing Standard, BigDye Terminator v1.1 Dx, consists of four tubes, each of which contains DNA of a known 1,200 base pair sequence prepared with BigDye Terminator v1.1. Sufficient DNA is included to perform one spectral calibration/performance check and/or one control sequencing run on an Applied Biosystems Dx genetic analyzer. The DNA preparation has been lyophilized to maximize stability.

Note: Applied Biosystems Dx genetic analyzers and consumables are for distribution and use in specific European countries only. Not for use in the USA.

This BigDye® Terminator v1.1 sequencing standard consists of four tubes, each of which contains DNA of a known 1,200 base pair sequence prepared with BigDye® Terminator v1.1.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye™ Terminator v1.1. Each tube can perform one spectral calibration/performance check and/or one control sequencing run on the Applied Biosystems 3500 Dx Series Genetic Analyzers. Store kit at -15 to -25°C.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Sequencing
Label or Dye ROX, TAMRA™, Rhodamine 110, Rhodamine 6G
Product Line BigDye™ Terminator
Product Type Sequencing Standard
Quantity 4 Tubes

For In Vitro Diagnostic Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.