Learn More
STEMCELL Technologies 35 mm Culture Dishes
Click to view available options
Quantity:
10 Dishes
500 Dishes
Unit Size:
Pack of 10
Pack of 500
Description
- 35 mm culture dishes, pre-tested for optimal colony growth without supporting anchorage-dependent cells in methylcellulose-based assays
- They are manufactured from optical quality, thermally resistant, non-pyrogenic, non-toxic polystyrene
- Recommended for: Hematopoietic progenitor cultures in methylcellulose-based media (MethoCult™)
- Embryoid body formation from mouse embryonic and induced pluripotent stem cells
- Performing the neural colony-forming cell assay (NCFC)
Specifications
Specifications
| For Use With (Application) | Cell Culture; Colony Assay |
| Quantity | 500 Dishes |