missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ BF4 Cotton Linter Blotting Paper
Shop All Sartorius Products
Click to view available options
Length (Metric):
580 mm
Unit Size:
Pack of 50
Description
- Made of high purity cotton linters for uniform buffer flow and resulting blots
- No additives to avoid interference during transfer
- Supplied in sheets
Specifications
Specifications
| Length (Metric) | 580 mm |
| Width (Metric) | 600 mm |
| Color | White |
| Quantity | 50/Pk. |
| Material | Cotton linters with an alpha-cellulose content |
| Thickness | 0.76 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction