missing translation for 'onlineSavingsMsg'
Learn More

33-O-Formyl Rapamycin (>85%), TRC

Product Code. 30395159
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395159 5 mg 5mg
30391014 1 mg 1mg
30407512 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395159 Supplier Toronto Research Chemicals Supplier No. TRCF6990105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 33-O-Formyl Rapamycin
Analyte or Component Names 33-O-Formyl Rapamycin
Shipping Condition Room Temperature
Molecular Formula C52 H79 N O14
Synonym 40-O-Formyl Rapamycin, 42-O-Formyl Rapamycin
InChI Formula InChI=1S/C52H79NO14/c1-31-16-12-11-13-17-32(2)43(62-8)28-39-21-19-37(7)52(61,67-39)49(58)50(59)53-23-15-14-18-40(53)51(60)66-44(34(4)26-38-20-22-42(65-30-54)45(27-38)63-9)29-41(55)33(3)25-36(6)47(57)48(64-10)46(56)35(5)24-31/h11-13,16-17,25,30-31,33-35,37-40,42-45,47-48,57,61H,14-15,18-24,26-29H2,1-10H3/b13-11+,16-12+,32-17+,36-25+/t31-,33?,34-,35-,37-,38?,39+,40+,42-,43+,44+,45-,47-,48+,52-/m1/s1
Solution Type Neat
SMILES CO[C@@H]1CC(C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(/C)\[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)OC)CC[C@H]1OC=O
IUPAC Name [(1R,2R)-4-[(2R)-2-[(1R,9S,12S,15R,16E,18R,19R,21R,23S,24E,26E,28E,30S,32S,35R)-1,18-dihydroxy-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-2,3,10,14,20-pentaoxo-11,36-dioxa-4-azatricyclo[30.3.1.04,9]hexatriaconta-16,24,26,28-tetraen-12-yl]propyl]-2-methoxycyclohexyl] formate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 942.182
Quantity 5 mg
Formula Weight 941.55
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.