missing translation for 'onlineSavingsMsg'
Learn More

Nalgene 32 oz. Narrow Mouth Bottle

Product Code. 17150056
Change view
Click to view available options
Color:
Blue
Gray
Seafoam
Trout Green
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17150056 Blue Each
17140056 Gray Each
17160056 Trout Green Each
17170056 Seafoam Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17150056 Supplier Nalgene Supplier No. 20782028
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

All the effectiveness of the wide mouth bottle with a narrower opening for controlled hydration.

  • Specially designed lip ideal for keeping the flow, without the stress of a mess
  • Tough and guaranteed for life
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 946 mL
Capacity (English) 32 oz.
Color Blue
Mouth Narrow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.