Learn More
Masterflex™ Reducing T-Connector, Barbed Fitting, 316 Stainless Steel
Description
- 316 stainless steel construction
- Ideal for low-pressure applications (generally less than 30psi)
Specifications
Specifications
| Material | Stainless Steel (316) |
| For Use With | 1/4 in. x 1/8 in. ID tubing |
| For Use With (Application) | Low-pressure applications |
| Packaging Quantity | 1 Each |
| Tubing Size | 1/4 in. x 1/8 in. ID tubing |
| Port Size | 3/32 in. orifice; 1-5/16 x 17/32 in. |
| Shape | T-shape |
| Product Type | Reducing connector |