Learn More
Masterflex™ 316 Stainless Steel NPT Male Pipe Adapter Barbed Fitting
Click to view available options
For Use With:
1/4 NPT(M) x 1/4 in. tubing I.D.
1/4 NPT(M) x 1/8 in. tubing I.D.
1/4 NPT(M) x 3/8 in. tubing I.D.
1/8 NPT(M) x 1/16 in. tubing I.D.
1/8 NPT(M) x 1/8 in. tubing I.D.
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel (316) |
| For Use With | 1/8 NPT(M) x 1/16 in. tubing I.D. |
| Packaging Quantity | 1 Each |
| Port Size | 1/16 in. orifice; 3/4 x 17/16 in. |
| Thread Size | 1/8 NPT(M) |
| Product Type | Barbed Fitting |