missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 310 Genetic Analyzer Matrix Standards Kit with Manual and Software, dRhodamine
Shop All Applied Biosystems Products
Click to view available options
Label or Dye:
dRhodamine 110, dRhodamine 6G, dRhodamine ROX, dRhodamine TAMRA
Unit Size:
Each
Description
Includes
Each kit contains a set of four standards used to generate a matrix file for the 310 Genetic Analyzer. Each tube contains a different fluorescent dye in solution with blue loading dye. Contents are stable for 6 months at 2 to 8°C. Includes Manual and Software
The kit contains matrix standards for use with dRhodamine sequencing chemistries on the 310 Genetic Analyzer.
Order Info
1 Kit
Specifications
Specifications
| Content And Storage | Each kit contains a set of four standards used to generate a matrix file for the 310 Genetic Analyzer. Each tube contains a different fluorescent dye in solution with blue loading dye. Contents are stable for 6 months at 2 to 8°C. Includes Manual and Software. |
| Format | Kit |
| For Use With (Application) | Sequencing |
| Label or Dye | dRhodamine 110, dRhodamine 6G, dRhodamine ROX, dRhodamine TAMRA |
| Product Type | 310 Genetic Analyzer Matrix Standards Kit |
| Quantity | 1 Kit |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction