missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRBP1 Antibody (OTI3F1), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44996-30ul
This item is not returnable.
View return policy
Description
RRBP1 Monoclonal antibody specifically detects RRBP1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| RRBP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| 6238 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI3F1 | |
| Western Blot 1:2000, Immunohistochemistry 1:1000 | |
| 180 kDa ribosome receptor homolog, DKFZp586A1420, ES/130, ES/130-related protein, ES130, FLJ36146, hES, KIAA1398, MGC157720, MGC157721, ribosome binding protein 1 (dog 180kD homolog), ribosome binding protein 1 homolog 180kDa (dog), Ribosome receptor protein, ribosome-binding protein 1, RRp | |
| Human recombinant protein fragment corresponding to amino acids 484-590 of human RRBP1(NP_001036041) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction