missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ H4K5ac Recombinant Rabbit Monoclonal Antibody (K01_2Q13)

Product Code. 30828111
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828111 50 μL
30830036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828111 Supplier Invitrogen™ Supplier No. MA565704
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

H4K5ac Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H4K5ac
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2Q13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene H4C1
Gene Accession No. P62805, P62806
Gene Alias acetyl Histone H4; Acetyl-H4 K5; Acetyl-H4 Lys5; Acetyl-Histone H4 K5; Acetyl-Histone H4 Lys5; B130044J01Rik; FO108; germinal histone H4; H1ft; H4; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4 K5ac; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/j; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4C1; H4C11; H4C12; H4C13; H4C14; H4C15; H4C2; H4c3; H4C4; H4C5; H4c6; H4c8; H4C9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4ft; H4K5; H4Lys5ac; H4M; HIST1H4; Hist1h4a; HIST1H4B; Hist1h4c; Hist1h4d; HIST1H4E; HIST1H4F; Hist1h4h; HIST1H4I; Hist1h4j; HIST1H4K; HIST1H4L; Hist1h4m; HIST2H4; HIST2H4A; HIST2H4B; Hist4; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; histone H4; Histone H4 acetyl; Histone H4A; Histone H4K5; Histone H4K5ac; histone IV, family 2; LOC680097; OGP; Osteogenic growth peptide; RGD1565073; similar to germinal histone H4 gene; X04652
Gene Symbols H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9
Host Species Rabbit
Immunogen A synthesized peptide derived from human Histone H4 (acetyl K5).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121504, 319158, 326619, 326620, 8294, 8359, 8363, 8366, 8367, 8370, 97122
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.