missing translation for 'onlineSavingsMsg'
Learn More

3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific™

A selective 5-HT3 serotonin receptor antagonist

£157.00

Chemical Identifiers

CAS 105826-92-4
Molecular Formula C17H21ClN2O2
Molecular Weight (g/mol) 320.817
MDL Number MFCD00210221
InChI Key XIEGSJAEZIGKSA-ICHLHHFLSA-N
Synonym sdz-ics 930
PubChem CID 71300439
IUPAC Name [(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride
SMILES CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15435109
SDS View Documents
Thermo Scientific Alfa Aesar
J62807.MC
100mg
£157.00
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

105826-92-4
320.817
XIEGSJAEZIGKSA-ICHLHHFLSA-N
71300439
CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl
C17H21ClN2O2
MFCD00210221
sdz-ics 930
[(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride

Specifications

105826-92-4
MFCD00210221
14,9783
Soluble in water
CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl
320.817
320.82
C17H21ClN2O2
UN2811
sdz-ics 930
XIEGSJAEZIGKSA-ICHLHHFLSA-N
[(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride
71300439
3-Tropanylindole-3-carboxylate hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'rtecsNumber' : NL6012520

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.