missing translation for 'onlineSavingsMsg'
Learn More

3-(Tritylthio)propionic acid, 97%

Product Code. 15446568
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446568 Supplier Thermo Scientific Alfa Aesar Supplier No. H60093.03
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27144-18-9 | C22H20O2S | 348.46 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Partly miscible in water.

Notes
Stor
TRUSTED_SUSTAINABILITY

Specifications

CAS 27144-18-9
Molecular Formula C22H20O2S
Linear Formula (C6H5)3CSCH2CH2COOH
MDL Number MFCD00237291
Quantity 1 g
Sensitivity Air Sensitive
Synonym 3-tritylthio propanoic acid, s-trityl-3-mercaptopropionic acid, 3-tritylthio propionic acid, 3-tritylsulfanylpropionic acid, 3-triphenylmethyl sulfanyl propanoic acid, 3-tritylsulfanyl-propionic acid, 3-tritylmercapto propionic acid, s-trityl-beta-mercaptopropionic acid, propanoic acid, 3-triphenylmethyl thio, 3-tritylsulfanyl propanoic acid
Solubility Information Partly miscible in water.
InChI Key AECGEIVNZGQBJT-UHFFFAOYSA-N
SMILES OC(=O)CCSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 3-tritylsulfanylpropanoic acid
Molecular Weight (g/mol) 348.46
PubChem CID 262767
Formula Weight 348.46
Percent Purity 97%
Chemical Name or Material 3-(Tritylthio)propionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.