missing translation for 'onlineSavingsMsg'
Learn More

3-Tritylsulfanylpropionic Acid, TRC

Product Code. 30452630
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452630 100 g -
30452423 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452630 Supplier Toronto Research Chemicals Supplier No. TRCT889475100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27144-18-9
Molecular Formula C22H20O2S
Molecular Weight (g/mol) 348.46
Synonym 3-[(Triphenylmethyl)thio]propanoic Acid, 3-(Triphenylmethylthio)propionic Acid, 3-(Tritylthio)propanoic Acid, 3-(Tritylthio)propionic Acid, 3-[(Triphenylmethyl)mercapto]propionic Acid, NSC 96707, S-Trityl-3-mercaptopropionic Acid, β-(Tritylmercapto)propionic Acid
IUPAC Name 3-tritylsulfanylpropanoic acid
SMILES OC(=O)CCSC(c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 100 g
InChI Formula InChI=1S/C22H20O2S/c23-21(24)16-17-25-22(18-10-4-1-5-11-18,19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15H,16-17H2,(H,23,24)
Formula Weight 348.1184
Chemical Name or Material 3-Tritylsulfanylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.