missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D

Product Code. 10535290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10535290 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10535290 Supplier Thermo Scientific Acros Supplier No. 432120010
  • ONLINE EXCLUSIVE PRICE
    £120.00
    £116.00 / 1g
    Save £4.00
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 24493-21-8 | C6H9D4NaO2Si | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

CAS 24493-21-8
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98 atom% D
Packaging Glass bottle
Molecular Formula C6H9D4NaO2Si
Linear Formula (CH3)3SiCD2CD2CO2Na
Quantity 1 g
Synonym sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate
Solubility Information Solubility in water: soluble
InChI Key OIIWPAYIXDCDNL-HGFPCDIYSA-M
SMILES C[Si](C)(C)CCC(=O)[O-].[Na+]
IUPAC Name sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate
Molecular Weight (g/mol) 172.27
PubChem CID 23688921
Formula Weight 172.27
Percent Purity 98 atom% D
Physical Form Powder
Chemical Name or Material 3-(Trimethylsilyl)propionic acid-d4 sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trimethylsilyl)propionic acid-d4 sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.