missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propiolic acid, 97%

Product Code. 15480138
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480138 25 g 25g
15460138 1 g 1g
15470138 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480138 Supplier Thermo Scientific Alfa Aesar Supplier No. H53380.14
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5683-31-8 | C6H10O2Si | 142.23 g/mol

3-(Trimethylsilyl)propiolic acid may be used in the regioselective preparation of 1,5-trisubstituted 1H-1,2,3-triazoles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trimethylsilyl)propiolic acid may be used in the regioselective preparation of 1,5-trisubstituted 1H-1,2,3-triazoles. It is also used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5683-31-8
Molecular Formula C6H10O2Si
Molecular Weight (g/mol) 142.23
MDL Number MFCD00190216
InChI Key IPEATTYBFBRNEB-UHFFFAOYSA-N
Synonym 3-trimethylsilyl propiolic acid, 3-trimethylsilyl propynoic acid, trimethylsilylpropiolic acid, 3-trimethylsilylpropynoic acid, 3-trimethylsilyl prop-2-ynoic acid, 2-propynoic acid, 3-trimethylsilyl, tms-propynoic acid, acmc-209lul, 3-trimethylsilyl propynoicacid, 3-trimethylsilyl propalic acid
PubChem CID 2760863
SMILES C[Si](C)(C)C#CC(O)=O

Specifications

Boiling Point 105°C to 110°C (10mmHg)
Melting Point 47°C to 49°C
Quantity 25 g
Linear Formula (CH3)3SiC≡CCO2H
Beilstein 1755674
Flash Point 96°C (205°F)
Solubility Information Soluble in methanol.
IUPAC Name 3-trimethylsilylprop-2-ynoic acid
Formula Weight 142.23
Percent Purity 97%
Chemical Name or Material 3-(Trimethylsilyl)propiolic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.