missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)-2-propynoic Acid Ethyl Ester, TRC

Product Code. 30450488
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450488 1 g 1g
30450128 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30450488 Supplier Toronto Research Chemicals Supplier No. TRCT7974901G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16205-84-8
Molecular Formula C8H14O2Si
Molecular Weight (g/mol) 170.28
Synonym Propiolic acid, (trimethylsilyl)-, ethyl ester (6CI,8CI), 1-(Ethoxycarbonyl)-2-(trimethylsilyl)acetylene, Ethyl (trimethylsilyl)ethynylcarboxylate, Ethyl (trimethylsilyl)propiolate, Ethyl 3-(trimethylsilyl)propiolate, Ethyl 3-(trimethylsilyl)propynoate, Ethyl 3-trimethylsilylprop-2-ynoate
IUPAC Name ethyl 3-trimethylsilylprop-2-ynoate
SMILES CCOC(=O)C#C[Si](C)(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H14O2Si/c1-5-10-8(9)6-7-11(2,3)4/h5H2,1-4H3
Formula Weight 170.0763
Chemical Name or Material 3-(Trimethylsilyl)-2-propynoic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.