missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethylthio)aniline, 96%

Product Code. 11400168
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400168 1 g 1g
11410168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400168 Supplier Thermo Scientific Alfa Aesar Supplier No. B23734.03
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 369-68-6 | C7H6F3NS | 193.19 g/mol

3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethylthio)aniline is used as an intermediate in organic synthesis. It is also involved in the regioselective Friedel-Crafts alkylation in hexafluoroacetone sesquihydrate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-68-6
Molecular Formula C7H6F3NS
Molecular Weight (g/mol) 193.19
MDL Number MFCD00190130
InChI Key DENPAKQJZNDKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethylthio aniline, 3-trifluoromethyl sulfanyl aniline, 3-aminophenyl trifluoromethyl sulphide, 3-aminophenyl trifluoromethyl sulfide, 3-trifluoromethyl thio aniline, 3-trifluoromethylsulfanyl aniline, benzenamine, 3-trifluoromethyl thio, benzeneamine, 3-trifluoromethyl thio, 3-trifluoromethylthio phenylamine
PubChem CID 2735939
IUPAC Name 3-(trifluoromethylsulfanyl)aniline
SMILES NC1=CC=CC(SC(F)(F)F)=C1

Specifications

Density 1.348
Boiling Point 221°C to 224°C
Flash Point 103°C (218°F)
Odor Unpleasant
Refractive Index 1.522
Quantity 1 g
UN Number UN2810
Solubility Information Immiscible with water.
Formula Weight 193.19
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethylthio)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.