missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethylbenzoic anhydride, 97%

Product Code. 15446637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446637 5 g 5g
15436637 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446637 Supplier Thermo Scientific Alfa Aesar Supplier No. H33779.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25753-15-5 | C16H8F6O3 | 362.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25753-15-5
Molecular Formula C16H8F6O3
Molecular Weight (g/mol) 362.23
MDL Number MFCD09757594
InChI Key BYLMYFXAIMTZIC-UHFFFAOYSA-N
Synonym 3-trifluoromethylbenzoic anhydride, 3-trifluoromethyl benzoic anhydride, benzoic acid, 3-trifluoromethyl-, anhydride, 3-trifluoromethyl benzoyl 3-trifluoromethyl benzoate, acmc-1cd8h, 3-trifluoromethyl phenyl anhydride #, benzoic acid,3-trifluoromethyl-, anhydride with 3-trifluoromethyl benzoic acid
PubChem CID 599431
IUPAC Name [3-(trifluoromethyl)benzoyl] 3-(trifluoromethyl)benzoate
SMILES FC(F)(F)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 362.23
Percent Purity 97%
Chemical Name or Material 3-Trifluoromethylbenzoic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.