missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethylbenzoic anhydride, 97%

Product Code. 15436637
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15436637

Brand: Thermo Scientific Alfa Aesar H33779.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25753-15-5 | C16H8F6O3 | 362.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25753-15-5
Molecular Formula C16H8F6O3
Molecular Weight (g/mol) 362.23
MDL Number MFCD09757594
InChI Key BYLMYFXAIMTZIC-UHFFFAOYSA-N
Synonym 3-trifluoromethylbenzoic anhydride, 3-trifluoromethyl benzoic anhydride, benzoic acid, 3-trifluoromethyl-, anhydride, 3-trifluoromethyl benzoyl 3-trifluoromethyl benzoate, acmc-1cd8h, 3-trifluoromethyl phenyl anhydride #, benzoic acid,3-trifluoromethyl-, anhydride with 3-trifluoromethyl benzoic acid
PubChem CID 599431
IUPAC Name [3-(trifluoromethyl)benzoyl] 3-(trifluoromethyl)benzoate
SMILES FC(F)(F)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Melting Point 74°C to 78°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 362.23
Percent Purity 97%
Chemical Name or Material 3-Trifluoromethylbenzoic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.