missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)styrene, 97+%, stabilized

Product Code. 10036122 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10036122 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10036122 Supplier Thermo Scientific Acros Supplier No. 388010010
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 402-24-4 | C9H7F3 | 172.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 402-24-4
CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 1.1610g/mL
Boiling Point 64.5°C (40.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Molecular Formula C9H7F3
Linear Formula CH2CHC6H4CF3
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
MDL Number MFCD00075481
Quantity 1 g
Specific Gravity 1.161
Synonym 3-trifluoromethyl styrene, 1-trifluoromethyl-3-vinylbenzene, 1-ethenyl-3-trifluoromethyl benzene, 3-cf3c6h4ch=ch2, 3-trifluoromethylstyren, 3-vinylbenzotrifluoride, m-trifluoromethylstyrene, pubchem15477, 3-trifluoromethylstyrene, 1-trifluoromethyl-3-vinyl-benzene
InChI Key ARHOUOIHKWELMD-UHFFFAOYSA-N
SMILES FC(F)(F)C1=CC=CC(C=C)=C1
IUPAC Name 1-ethenyl-3-(trifluoromethyl)benzene
Stabilizer about 100ppm 4-tert.-butylcatechol
Molecular Weight (g/mol) 172.15
PubChem CID 520986
Formula Weight 172.15
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)styrene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-(Trifluoromethyl)styrene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.