missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)salicylaldehyde, 98+%

Product Code. 15495617
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495617 250 mg 250mg
15485617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495617 Supplier Thermo Scientific Alfa Aesar Supplier No. H32858.MD
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 336628-67-2 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336628-67-2
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD03788529
InChI Key ZUNOXMJBNMKYOM-UHFFFAOYSA-N
Synonym 2-hydroxy-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl salicylaldehyde, 2-hydroxy-3-trifluoromethyl-benzaldehyde, benzaldehyde, 2-hydroxy-3-trifluoromethyl, pubchem10111, 2-oxidanyl-3-trifluoromethyl benzaldehyde
PubChem CID 2779274
IUPAC Name 2-hydroxy-3-(trifluoromethyl)benzaldehyde
SMILES OC1=C(C=O)C=CC=C1C(F)(F)F

Specifications

Melting Point 54°C to 59°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 190.12
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)salicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.