missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)piperidine, 97%

Product Code. 15463317
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463317 250 mg 250mg
11435590 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463317 Supplier Thermo Scientific Alfa Aesar Supplier No. H25851.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 768-31-0 | C6H11F3N | 154.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-31-0
Molecular Formula C6H11F3N
Molecular Weight (g/mol) 154.16
MDL Number MFCD02183561
InChI Key JOHFJTBDUSVGQB-RXMQYKEDSA-O
PubChem CID 2760776
SMILES FC(F)(F)[C@@H]1CCC[NH2+]C1

Specifications

Density 1.16
Boiling Point 128°C to 130°C
Flash Point 28°C (83°F)
Odor Amine-like
Refractive Index 1.393
Quantity 250 mg
UN Number UN2733
Sensitivity Air Sensitive
Synonym 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl
IUPAC Name (3R)-3-(trifluoromethyl)piperidin-1-ium
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)piperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.