missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide, 5% w/v in methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
25 mL
Description
Transesterification reactions.3-(Trifluoromethyl)phenyltrimethylammonium hydroxide is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 mL |
UN Number | UN2924 |
Solubility Information | Miscible with water and alcohols. |
Formula Weight | 221.22 |
Concentration or Composition (by Analyte or Components) | 5% w/v in methanol |
Physical Form | Liquid |
Chemical Name or Material | 3- (Trifluoromethyl)phenyltrimethylammonium hydroxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'