missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylhydrazine, 95%

Product Code. 11398545
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398545 1 g 1g
11308555 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398545 Supplier Thermo Scientific Alfa Aesar Supplier No. L11996.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 368-78-5 | C7H7F3N2 | 176.142 g/mol

3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethyl)phenylhydrazine is used in the synthesis of N-2-diphenylhydrazinecarboxamides and N-2-diphenylhydrazine carbothioamides.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-78-5
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00025093
InChI Key RSESUCWJKLHXEZ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylhydrazine, 3-trifluoromethyl phenyl hydrazine, 3-trifluoromethylphenylhydrazine, hydrazine, 3-trifluoromethyl phenyl, 3-trifloromethylphenylhydrazine, 3-trifluouromethyl phenylhydrazinehydrochloride, 3-trifluoromethyl phenylhydrazine hydrochioride, pubchem3280, enamine_001261
PubChem CID 98650
IUPAC Name [3-(trifluoromethyl)phenyl]hydrazine
SMILES C1=CC(=CC(=C1)NN)C(F)(F)F

Specifications

Density 1.348
Boiling Point 80°C to 83°C (9 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.504
Quantity 1 g
Beilstein 640075
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 176.14
Percent Purity 95%
Chemical Name or Material 3-(Trifluoromethyl)phenylhydrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.