missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylacetone, 97%, may cont. up to ca 5% monohydrate, Thermo Scientific™

Product Code. 11456170
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456170 25 g 25g
11446170 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456170 Supplier Thermo Scientific Alfa Aesar Supplier No. H27095.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21906-39-8
Molecular Formula C10H9F3O
Molecular Weight (g/mol) 202.176
MDL Number MFCD00000397
InChI Key JPHQCDCEBDRIOL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl propan-2-one, m-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl acetone, 3-trifluormethyl phenylacetone, 1-3-trifluoromethyl phenyl-2-propanone, m-trifluoromethylphenylacetone, 2-propanone, 1-3-trifluoromethyl phenyl
PubChem CID 89101
IUPAC Name 1-[3-(trifluoromethyl)phenyl]propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)C(F)(F)F

Specifications

Density 1.204
Boiling Point 89°C to 90°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.458
Quantity 25 g
Formula Weight 202.18
Percent Purity 97%
Assay may contain up to ≈5% monohydrate
Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.