missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenylacetic acid, 96%

Product Code. 11373096
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373096 50 g 50g
11363096 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373096 Supplier Thermo Scientific Alfa Aesar Supplier No. L10960.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 351-35-9 | C9H7F3O2 | 204.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351-35-9
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00004339
InChI Key BLXXCCIBGGBDHI-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetic acid, m-trifluoromethyl phenylacetic acid, 2-3-trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3-trifluoromethyl, 3-trifluoromethylphenylacetic acid, 3-trifluoromethyl phenyl acetic acid, a,a,a-trifluoro-m-tolyl acetic acid, alpha,alpha,alpha-trifluoro-m-tolyl acetic acid, 3-carboxymethyl benzotrifluoride
PubChem CID 67695
IUPAC Name 2-[3-(trifluoromethyl)phenyl]acetic acid
SMILES C1=CC(=CC(=C1)C(F)(F)F)CC(=O)O

Specifications

Melting Point 77°C to 78°C
Quantity 50 g
Beilstein 2213223
Formula Weight 204.15
Percent Purity ≥98%
Chemical Name or Material 3-(Trifluoromethyl)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.