missing translation for 'onlineSavingsMsg'
Learn More

[3-(Trifluoromethyl)phenyl]methanesulfonyl chloride, 90%, Thermo Scientific™

Catalog Number p-3075153
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
127162-96-3
C8H6ClF3O2S
258.639
MFCD01631926
WIGCBGFBBRJTLS-UHFFFAOYSA-N
3-trifluoromethylbenzylsulfonyl chloride, 3-trifluoromethyl phenyl methanesulfonyl chloride, 3-trifluoromethylbenzylsulfonylchloride, 3-trifluoromethyl phenyl methyl sulfonyl chloride, 3-chlorosulphonyl methyl benzotrifluoride, 3-trifluoromethyl phenylmethanesulfonyl chloride, 3-trifluoromethyl phenyl methylsulphonyl chloride, 3-trifluoromethyl benzylsulphonyl chloride, 3-trifluoromethyl phenyl methyl sulfonylchloride
2794646
[3-(trifluoromethyl)phenyl]methanesulfonyl chloride
C1=CC(=CC(=C1)C(F)(F)F)CS(=O)(=O)Cl
This item is not returnable. View return policy
127162-96-3
C8H6ClF3O2S
258.639
MFCD01631926
WIGCBGFBBRJTLS-UHFFFAOYSA-N
3-trifluoromethylbenzylsulfonyl chloride, 3-trifluoromethyl phenyl methanesulfonyl chloride, 3-trifluoromethylbenzylsulfonylchloride, 3-trifluoromethyl phenyl methyl sulfonyl chloride, 3-chlorosulphonyl methyl benzotrifluoride, 3-trifluoromethyl phenylmethanesulfonyl chloride, 3-trifluoromethyl phenyl methylsulphonyl chloride, 3-trifluoromethyl benzylsulphonyl chloride, 3-trifluoromethyl phenyl methyl sulfonylchloride
2794646
[3-(trifluoromethyl)phenyl]methanesulfonyl chloride
C1=CC(=CC(=C1)C(F)(F)F)CS(=O)(=O)Cl
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 258.65
Percent Purity 90%
Chemical Name or Material [3-(Trifluoromethyl)phenyl]methanesulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'