missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)isoxazole-4-carboxylic Acid, TRC

Product Code. 30455988
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455988 100 mg 100mg
30456648 10 mg -
30457229 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455988 Supplier Toronto Research Chemicals Supplier No. TRCT897310100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076245-98-1
Molecular Formula C5H2NO3F3
Molecular Weight (g/mol) 181.07
IUPAC Name 3-(trifluoromethyl)-1,2-oxazole-4-carboxylic acid
SMILES OC(=O)c1conc1C(F)(F)F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H2F3NO3/c6-5(7,8)3-2(4(10)11)1-12-9-3/h1H,(H,10,11)
Formula Weight 180.9987
Chemical Name or Material 3-(Trifluoromethyl)isoxazole-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.