missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15289656 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15289656 1 g 1g
15299656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15289656 Supplier Thermo Scientific Acros Supplier No. 450510010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 885267-96-9
Molecular Formula C13H8ClF3O2S
Molecular Weight (g/mol) 320.71
MDL Number MFCD06409268
InChI Key WJSSWLPZDVPLIK-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl biphenyl-4-sulfonyl chloride, 4-3-trifluoromethyl phenyl benzenesulfonyl chloride, 3'-trifluoromethyl-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-trifluoromethyl, 3'-trifluoromethyl 1,1'-biphenyl-4-sulfonyl chloride, 3'-trifluoromethylbiphenyl-4-sulfonyl chloride
PubChem CID 16244690
IUPAC Name 4-[3-(trifluoromethyl)phenyl]benzenesulfonyl chloride
SMILES FC(F)(F)C1=CC=CC(=C1)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 78°C to 82°C
Assay Percent Range 96% min. diethylsulfonamide derivative (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 320.72
Percent Purity 97%
Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.