missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzylamine, 97%

Product Code. 11441157
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441157 5 g 5g
11451157 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441157 Supplier Thermo Scientific Alfa Aesar Supplier No. B24143.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2740-83-2 | C8H8F3N | 175.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2740-83-2
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD00008117
InChI Key YKNZTUQUXUXTLE-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzylamine, 3-trifluoromethyl phenyl methanamine, 3-trifluoromethylbenzylamine, m-trifluoromethylbenzylamine, m-trifluoromethylbenzyl amine, 3-trifluoromethyl benzyl amine, benzenemethanamine, 3-trifluoromethyl, 3-trifluoromethyl-benzylamine, m-trifluoromethyl benzylamine
PubChem CID 75962
IUPAC Name [3-(trifluoromethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)C(F)(F)F)CN

Specifications

Density 1.222
Boiling Point 93°C to 97°C (28 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.463
Quantity 5 g
UN Number UN2735
Beilstein 1102465
Sensitivity Air Sensitive
Formula Weight 175.15
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.