missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzyl bromide, 98%

Product Code. 11419586
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11419586 1 g 1g
11429586 5 g 5g
11439586 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11419586

Brand: Thermo Scientific Alfa Aesar A19330.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 402-23-3 | C8H6BrF3 | 239.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-23-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.04
MDL Number MFCD00000395
InChI Key MYYYZNVAUZVXBO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzyl bromide, 1-bromomethyl-3-trifluoromethyl benzene, m-trifluoromethylbenzyl bromide, 3-trifluoromethylbenzyl bromide, 3-bromomethyl benzotrifluoride, 3-trifluoromethylbenzylbromide, 3-trifluoromethyl benzylbromide, 1-bromomethyl-3-trifluoromethyl-benzene, alpha'-bromo-alpha,alpha,alpha-trifluoro-m-xylene
PubChem CID 123061
IUPAC Name 1-(bromomethyl)-3-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC(CBr)=C1

Specifications

Boiling Point 67°C to 69°C (4 mmHg)
Flash Point 87°C (188°F)
Odor Pungent
Quantity 1 g
UN Number UN3265
Beilstein 743243
Formula Weight 239.04
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.