missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoyl chloride, 98%

Product Code. 10177603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177603 25 mL Glass bottle 25mL
10626681 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177603 Supplier Thermo Scientific Acros Supplier No. 312940250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2251-65-2 | C8H4ClF3O | 208.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2251-65-2
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.57
MDL Number MFCD00000680
InChI Key RUJYJCANMOTJMO-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-trifluoromethyl, m-trifluoromethyl benzoyl chloride, m-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-m-toluoyl chloride, 3-trifluoromethyl benzoylchloride, 3-trifluoromethyl-benzoyl chloride, alpha,alpha,alpha-trifluoro-m-tolyl chloride, pubchem5468
PubChem CID 75257
IUPAC Name 3-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.3800g/mL
Boiling Point 184.0°C to 186.0°C (750.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4COCl
Refractive Index 1.4760 to 1.4780
Quantity 25 mL
Specific Gravity 1.38
Solubility Information Solubility in water: decomposes
Formula Weight 208.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)benzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.