missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzoic acid, Thermo Scientific™

Product Code. 10285514
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10285514 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10285514 Supplier Thermo Scientific Maybridge Supplier No. DP01925EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 454-92-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H5F3O2
Linear Formula CF3C6H4CO2H
MDL Number MFCD00002519
Quantity 10g
Synonym 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid
InChI Key FQXQBFUUVCDIRK-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC(=C1)C(F)(F)F
IUPAC Name 3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 190.12
PubChem CID 9963
ChEBI CHEBI:60695
Formula Weight 190.12
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.