missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzenesulfonyl chloride, 98%

Product Code. 11339549
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339549 1 g 1g
11417626 5 g 5g
11427626 25 g 25g
11437626 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11339549 Supplier Thermo Scientific Alfa Aesar Supplier No. A14657.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 777-44-6 | C7H4ClF3O2S | 244.612 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 777-44-6
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.612
MDL Number MFCD00014724
InChI Key ONCAZCNPWWQQMW-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
PubChem CID 2733250
IUPAC Name 3-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F

Specifications

Density 1.526
Boiling Point 79°C to 80°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.487
Quantity 1 g
UN Number UN3265
Beilstein 2650774
Sensitivity Moisture sensitive
Formula Weight 244.62
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.