missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)benzeneboronic acid, 98%

Product Code. 11490227
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490227 1 g 1g
11400237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490227 Supplier Thermo Scientific Alfa Aesar Supplier No. B21661.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1423-26-3 | C7H6BF3O2 | 189.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1423-26-3
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
MDL Number MFCD00151854
InChI Key WOAORAPRPVIATR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
PubChem CID 2734388
IUPAC Name [3-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 162°C to 166°C
Quantity 1 g
Beilstein 6084746
Formula Weight 189.93
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.