missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)anisole, 99%, Thermo Scientific™

Product Code. 10501841 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10501841

Brand: Thermo Scientific Acros 199780050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 454-90-0
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.2100g/mL
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H7F3O
Linear Formula CF3C6H4OCH3
Refractive Index 1.4424 to 1.4444
MDL Number MFCD00000385
Quantity 5 g
Specific Gravity 1.21
Synonym 3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
InChI Key XHONYVFDZSPELQ-UHFFFAOYSA-N
SMILES COC1=CC=CC(=C1)C(F)(F)F
IUPAC Name 1-methoxy-3-(trifluoromethyl)benzene
Molecular Weight (g/mol) 176.14
PubChem CID 96691
Formula Weight 176.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)anisole
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trifluoromethyl)anisole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.