missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)anisole, 98%, Thermo Scientific Chemicals

Product Code. 11440788
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440788 25 g 25g
11430788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440788 Supplier Thermo Scientific Alfa Aesar Supplier No. B25249.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 454-90-0 | Molecular Formula: C8H7F3O | Molecular Weight: 176.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-90-0
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.14
MDL Number MFCD00000385
InChI Key XHONYVFDZSPELQ-UHFFFAOYSA-N
Synonym 3-trifluoromethyl anisole, 1-methoxy-3-trifluoromethyl benzene, 3-methoxybenzotrifluoride, 3-trifluoromethylanisole, benzene, 1-methoxy-3-trifluoromethyl, 3-methoxy-1-trifluoromethyl benzene, pubchem2712, 3-trifluoromethylanisol, acmc-1cuj0, m-trifluoromethyl anisole
PubChem CID 96691
IUPAC Name 1-methoxy-3-(trifluoromethyl)benzene
SMILES COC1=CC=CC(=C1)C(F)(F)F

Specifications

Density 1.215
Boiling Point 159°C to 160°C
Flash Point 48°C (118°F)
Refractive Index 1.4435
Quantity 25 g
UN Number UN1993
Beilstein 3048340
Formula Weight 176.14
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethyl)anisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.