missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethyl)acetophenone, 99%, Thermo Scientific™

Product Code. 10703221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10703221 1 g Glass bottle 1g
10032950 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10703221 Supplier Thermo Scientific Acros Supplier No. 214440010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-76-8
MDL Number MFCD00000391
InChI Key ABXGMGUHGLQMAW-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl acetophenone, 3-trifluoromethyl acetophenone, 3-trifluoromethylacetophenone, ethanone, 1-3-trifluoromethyl phenyl, 1-3-trifluoromethyl phenyl ethanone, m-trifluoromethylacetophenone, 3'-trifluoromethylacetophenone, 1-3-trifluoromethyl phenyl ethan-1-one, 1-3-trifluoromethyl-phenyl-ethanone
PubChem CID 67682
IUPAC Name 1-[3-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Density 1.2300g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H7F3O
Linear Formula CF3C6H4COCH3
Refractive Index 1.4610 to 1.4640 (20°C, 589nm)
Quantity 1 g
Beilstein 07, III, 1057
Fieser 13,226
Molecular Weight (g/mol) 188.149
Formula Weight 188.15
Percent Purity ≥98.5% (GC)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3'-(Trifluoromethyl)acetophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.