missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethyl)acetanilide, 98+%, Thermo Scientific™

Product Code. 11403127
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403127 10 g 10g
11413127 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403127 Supplier Thermo Scientific Alfa Aesar Supplier No. A16098.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351-36-0
Molecular Formula C9H8F3NO
Molecular Weight (g/mol) 203.164
MDL Number MFCD00000383
InChI Key HNIPNANLYHXYDE-UHFFFAOYSA-N
Synonym 3-trifluoromethyl acetanilide, 3-acetamidobenzotrifluoride, m-trifluoromethylacetanilide, m-acetaminobenzotrifluoride, 3'-trifluoromethyl acetanilide, 3-trifluoromethylacetanilide, n-3-trifluoromethyl phenyl acetamide, usaf ma-14, acetanilide, 3-trifluoromethyl, 3'-trifluoromethylacetanilide
PubChem CID 9595
IUPAC Name N-[3-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 103°C to 106°C
Odor Odorless
Quantity 10 g
Beilstein 2213222
Formula Weight 203.16
Percent Purity ≥98%
Chemical Name or Material 3'-(Trifluoromethyl)acetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.