missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine Hydrochloride, TRC

Product Code. 30452092
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452092 5 g 5g
30452334 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452092 Supplier Toronto Research Chemicals Supplier No. TRCT7918105G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
CAS 762240-92-6
Molecular Formula C6 H7 F3 N4 . Cl H
Quantity 5 g
Synonym 1,2,4-Triazolo[4,3-a]pyrazine','5,6,7,8-tetrahydro-3-(trifluoromethyl)-','hydrochloride (1:1), 1,2,4-Triazolo[4,3-a]pyrazine','5,6,7,8-tetrahydro-3-(trifluoromethyl)-','monohydrochloride (9CI), 3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride, 3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine monohydrochloride
InChI Formula InChI=1S/C6H7F3N4.ClH/c7-6(8,9)5-12-11-4-3-10-1-2-13(4)5;/h10H,1-3H2;1H
SMILES Cl.FC(F)(F)c1nnc2CNCCn12
IUPAC Name 3-(trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine;hydrochloride
Molecular Weight (g/mol) 228.6
Formula Weight 228.039
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.