missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)-2-picolinic Acid, TRC

Product Code. 30452465
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
This item is not returnable. View return policy

Product Code. 30452465

Brand: Toronto Research Chemicals TRCT7906802.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Trifluoromethyl)-2-picolinic Acid
CAS 87407-12-3
Molecular Formula C7H4F3NO2
Quantity 2.5 g
Synonym 3-(Trifluoromethyl)-2-pyridinecarboxylic Acid, 3-Trifluoromethylpyridine-2-carboxylic Acid
InChI Formula InChI=1S/C7H4F3NO2/c8-7(9,10)4-2-1-3-11-5(4)6(12)13/h1-3H,(H,12,13)
SMILES OC(=O)c1ncccc1C(F)(F)F
IUPAC Name 3-(trifluoromethyl)pyridine-2-carboxylic acid
Molecular Weight (g/mol) 191.11
Formula Weight 191.0194
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.