missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethyl-1H-pyrazole, 97%

Product Code. 15491198
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491198 250 mg 250mg
15471198 1 g 1g
15481198 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491198 Supplier Thermo Scientific Alfa Aesar Supplier No. H55068.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20154-03-4 | C4H3F3N2 | 136.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20154-03-4
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.077
MDL Number MFCD00115018
InChI Key PYXNITNKYBLBMW-UHFFFAOYSA-N
Synonym 3-trifluoromethyl pyrazole, 3-trifluoromethyl-1h-pyrazole, 5-trifluoromethyl-1h-pyrazole, 3-trifluoromethylpyrazole, trifluoromethylpyrazole, 1h-pyrazole, 3-trifluoromethyl, 3-trifluoromethyl-2h-pyrazole, pubchem10164, pubchem10423
PubChem CID 1807034
IUPAC Name 5-(trifluoromethyl)-1H-pyrazole
SMILES C1=C(NN=C1)C(F)(F)F

Specifications

Melting Point 45°C to 47°C
Boiling Point 70°C (2 mmHg)
Quantity 250 mg
Beilstein 1564179
Formula Weight 136.08
Percent Purity 97%
Chemical Name or Material 3-Trifluoromethyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.