missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11429556
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429556 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11429556 Supplier Thermo Scientific Alfa Aesar Supplier No. A19296.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52771-21-8
Density 1.33
Boiling Point 83°C to 86°C (24 mmHg)
Flash Point 72°C (161°F)
Molecular Formula C8H5F3O2
Refractive Index 1.454
MDL Number MFCD00042406
Quantity 10 g
Beilstein 7267831
Sensitivity Air Sensitive
Synonym 3-trifluoromethoxy benzaldehyde, m-trifluoromethoxy benzaldehyde, m-trifluoromethoxybenzaldehyde, pubchem1470, acmc-209l1m, 3trifluoromethoxybenzaldehyde, 3-trifluormethoxy-benzaldehyde, ksc274m0j, 3-trifluoromethoxy-benzaldehyde, timtec-bb sbb006601
InChI Key FQEVHRCPXFKJHF-UHFFFAOYSA-N
SMILES FC(F)(F)OC1=CC=CC(C=O)=C1
IUPAC Name 3-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol) 190.12
PubChem CID 605139
Formula Weight 190.12
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethoxy)benzaldehyde
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.