missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)aniline, 98%

Product Code. 11434147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434147 1 g 1g
11444147 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434147 Supplier Thermo Scientific Alfa Aesar Supplier No. A19592.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1535-73-5 | C7H6F3NO | 177.126 g/mol

3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1535-73-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00041511
InChI Key SADHVOSOZBAAGL-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy aniline, 3-trifluoromethoxy-phenylamine, m-aminophenyl trifluoromethyl ether, m-trifluoromethoxy aniline, benzenamine, 3-trifluoromethoxy, alpha,alpha,alpha-trifluoro-m-anisidine, 3-trifluoromethoxy benzenamine, 3-trifluoromethoxy phenylamine, pubchem4507
PubChem CID 73753
IUPAC Name 3-(trifluoromethoxy)aniline
SMILES C1=CC(=CC(=C1)OC(F)(F)F)N

Specifications

Density 1.325
Boiling Point 72°C to 75°C (8 mmHg)
Flash Point 85°C (185°F)
Odor Characteristic
Refractive Index 1.466
Quantity 1 g
UN Number UN2810
Beilstein 776921
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 177.13
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.