missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenesulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10394082
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10394082 250 mg 250mg
10623783 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10394082 Supplier Thermo Scientific Maybridge Supplier No. CC13103CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 51175-71-4
Molecular Formula C4H3ClO2S2
Molecular Weight (g/mol) 182.64
MDL Number MFCD02682011
InChI Key YSPWSQNKRBSICH-UHFFFAOYSA-N
Synonym 3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride, chloro-3-thienylsulfone, acmc-209ksa, 3-thienylsulfonyl chloride, 3-thiophenesulphonylchloride, 3-thienyl-sulfonyl chloride, 3-chlorosulphonyl thiophene, 3-thienyl sulfonyl chloride, 3-thiophene sulfonyl chloride
PubChem CID 2776378
IUPAC Name thiophene-3-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CSC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 182.65
Percent Purity 97%
Chemical Name or Material 3-Thiophenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.