missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenecarboxylic Acid, TRC

Product Code. 30450328
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450328 25 g 25g
30450702 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30450328 Supplier Toronto Research Chemicals Supplier No. TRCT34483525G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiophene-3-carboxylic Acid
CAS 88-13-1
Molecular Formula C5 H4 O2 S
Quantity 25 g
Synonym 3-Thiophenecarboxylic acid, 3-Carboxythiophene, 3-Thenoic acid, 3-Thienylcarboxylic acid, NSC 66314, β-Thiophenecarboxylic acid, β-Thiophenic acid
InChI Formula InChI=1S/C5H4O2S/c6-5(7)4-1-2-8-3-4/h1-3H,(H,6,7)
SMILES OC(=O)c1ccsc1
IUPAC Name thiophene-3-carboxylic acid
Molecular Weight (g/mol) 128.15
Formula Weight 127.9932
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.