missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenecarboxaldehyde, 98%

Product Code. 10051911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051911 5 mL Glass bottle 5mL
10216150 25 mL Glass bottle 25mL
15149223 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051911 Supplier Thermo Scientific Acros Supplier No. 187980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 498-62-4 | C5H4OS | 112.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-62-4
MDL Number MFCD00005466
InChI Key RBIGKSZIQCTIJF-UHFFFAOYSA-N
Synonym 3-thiophenecarboxaldehyde, thiophene-3-carboxaldehyde, 3-formylthiophene, 3-thenaldehyde, 3-formyl-thiophene, 3-thiophenealdehyde, thiophene-3-aldehyde, 3-thiophene carboxaldehyde, 3-thiophenecarbaldehyde, 3-thiophenaldehyde
PubChem CID 68135
ChEBI CHEBI:87611
IUPAC Name thiophene-3-carbaldehyde
SMILES C1=CSC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -30.0°C
Color Yellow to Brown
Density 1.2800g/mL
Boiling Point 194.0°C to 196.0°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H4OS
Refractive Index 1.5820 to 1.5860
Quantity 5 mL
Beilstein 17,III,4497
Specific Gravity 1.28
Molecular Weight (g/mol) 112.15
Formula Weight 112.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Thiophenecarboxaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Thiophenecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.