missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophenecarbonyl chloride, Maybridge

Product Code. 10466842
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10466842 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10466842 Supplier Thermo Scientific Maybridge Supplier No. CC13102DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 41507-35-1
Assay Percent Range 97%
Molecular Formula C5H3ClOS
MDL Number MFCD00130090
Quantity 1 g
Synonym 3-thiophenecarbonyl chloride, 3-thenoyl chloride, 3-thiophenecarboxylic acid chloride, 3-thiophenecarbonylchloride, pubchem5597, 3-chlorocarbonyl thiophene, 3-thiophene carbonyl chloride, thiophene-3-carboxylic acid chloride, thiophene-3-carbonyl chloride, 3-thiophenecarbonyl chloride 6ci, 9ci
InChI Key QTWBEVAYYDZLQL-UHFFFAOYSA-N
SMILES ClC(=O)C1=CSC=C1
IUPAC Name thiophene-3-carbonyl chloride
Molecular Weight (g/mol) 146.59
PubChem CID 2776377
Formula Weight 146.6
Percent Purity 97%
Chemical Name or Material 3-Thiophenecarbonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.